Product Details
- SNP ID
-
rs183446770
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:38115563 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTCCTGATTGTACTCATGGATCTC[A/G]GTCATGGCATCCAGCGTGGGGTTGT
- Phenotype
-
MIM: 603604
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BAIAP2L2
PubMed Links
Gene Details
- Gene
- BAIAP2L2
- Gene Name
- BAI1 associated protein 2 like 2
There are no transcripts associated with this gene.
- Gene
- PLA2G6
- Gene Name
- phospholipase A2 group VI
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004426.1 |
2183 |
Silent Mutation |
ACC,ACT |
T612T |
NP_001004426.1 |
NM_001199562.1 |
2183 |
Silent Mutation |
ACC,ACT |
T612T |
NP_001186491.1 |
NM_003560.2 |
2183 |
Silent Mutation |
ACC,ACT |
T666T |
NP_003551.2 |
XM_005261764.2 |
2183 |
Silent Mutation |
ACC,ACT |
T666T |
XP_005261821.1 |
XM_005261765.1 |
2183 |
Silent Mutation |
ACC,ACT |
T666T |
XP_005261822.1 |
XM_005261766.1 |
2183 |
Silent Mutation |
ACC,ACT |
T666T |
XP_005261823.1 |
XM_006724332.3 |
2183 |
Silent Mutation |
ACC,ACT |
T666T |
XP_006724395.1 |
XM_011530422.1 |
2183 |
Silent Mutation |
ACC,ACT |
T631T |
XP_011528724.1 |
XM_011530423.1 |
2183 |
Silent Mutation |
ACC,ACT |
T488T |
XP_011528725.1 |
XM_011530424.1 |
2183 |
Silent Mutation |
ACC,ACT |
T488T |
XP_011528726.1 |
XM_011530425.1 |
2183 |
Silent Mutation |
ACC,ACT |
T488T |
XP_011528727.1 |
XM_011530426.2 |
2183 |
Intron |
|
|
XP_011528728.1 |
XM_017028981.1 |
2183 |
Silent Mutation |
ACC,ACT |
T612T |
XP_016884470.1 |
XM_017028982.1 |
2183 |
Silent Mutation |
ACC,ACT |
T612T |
XP_016884471.1 |
XM_017028983.1 |
2183 |
Silent Mutation |
ACC,ACT |
T434T |
XP_016884472.1 |
XM_017028984.1 |
2183 |
Silent Mutation |
ACC,ACT |
T434T |
XP_016884473.1 |
XM_017028985.1 |
2183 |
Silent Mutation |
ACC,ACT |
T440T |
XP_016884474.1 |
XM_017028986.1 |
2183 |
Intron |
|
|
XP_016884475.1 |
XM_017028987.1 |
2183 |
Intron |
|
|
XP_016884476.1 |
XM_017028988.1 |
2183 |
Intron |
|
|
XP_016884477.1 |
View Full Product Details