Product Details
- SNP ID
-
rs184115758
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:28495647 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGGGCGGAGTCAGAGTGGACCTG[C/G]CATGGGGAGACGGAGGGGAAGGCTG
- Phenotype
-
MIM: 605220
MIM: 607042
MIM: 608273
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
APOBR
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs149271,rs28576169] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- APOBR
- Gene Name
- apolipoprotein B receptor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018690.3 |
639 |
Missense Mutation |
TGC,TGG |
C202W |
NP_061160.3 |
- Gene
- CLN3
- Gene Name
- CLN3, battenin
There are no transcripts associated with this gene.
- Gene
- IL27
- Gene Name
- interleukin 27
There are no transcripts associated with this gene.
View Full Product Details