Product Details

SNP ID
rs184337874
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:62580639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAGAGCATTACGTTATATTAAC[A/G]TTACAAATTGTGTTACATCTTCAAT
Phenotype
MIM: 610889
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IPO11 PubMed Links

Gene Details

Gene
IPO11
Gene Name
importin 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134779.1 1440 Intron NP_001128251.1
NM_016338.4 1440 Intron NP_057422.3
Gene
IPO11-LRRC70
Gene Name
IPO11-LRRC70 readthrough
There are no transcripts associated with this gene.

Gene
LRRC70
Gene Name
leucine rich repeat containing 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181506.4 1440 Missense Mutation ATT,GTT I401V NP_852607.3

View Full Product Details