Product Details

SNP ID
rs184506462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33202377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGACATGGTCACAGTCATGGAC[A/G]TGGACACGCTCACAGTCATACACGT
Phenotype
MIM: 120290 MIM: 601417 MIM: 611500 MIM: 602045 MIM: 180246 MIM: 601416
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COL11A2 PubMed Links

Gene Details

Gene
COL11A2
Gene Name
collagen type XI alpha 2
There are no transcripts associated with this gene.

Gene
HSD17B8
Gene Name
hydroxysteroid 17-beta dehydrogenase 8
There are no transcripts associated with this gene.

Gene
MIR219A1
Gene Name
microRNA 219a-1
There are no transcripts associated with this gene.

Gene
RING1
Gene Name
ring finger protein 1
There are no transcripts associated with this gene.

Gene
RXRB
Gene Name
retinoid X receptor beta
There are no transcripts associated with this gene.

Gene
SLC39A7
Gene Name
solute carrier family 39 member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077516.1 1169 Missense Mutation CAT,CGT H250R NP_001070984.1
NM_001288777.1 1169 Missense Mutation CAT,CGT H125R NP_001275706.1
NM_006979.2 1169 Missense Mutation CAT,CGT H250R NP_008910.2

View Full Product Details