Product Details

SNP ID
rs184508752
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:121727852 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCATCTTTATCATCATCATTCA[G/T]TTCAGCCACTTGAGATTTTCGCCTC
Phenotype
MIM: 611970
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NIFK PubMed Links

Gene Details

Gene
NIFK
Gene Name
nucleolar protein interacting with the FHA domain of MKI67
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032390.4 831 Missense Mutation ATG,CTG M252L NP_115766.3
Gene
NIFK-AS1
Gene Name
NIFK antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details