Product Details

SNP ID
rs184482815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17436612 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCTCTCCCAGCCTCCCTCCAGG[A/G]CTGCAGGTGCTGTGGCAGTGGCCAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM221 PubMed Links

Gene Details

Gene
TMEM221
Gene Name
transmembrane protein 221
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190844.1 827 Missense Mutation GCC,GTC A241V NP_001177773.1
XM_011527603.2 827 Intron XP_011525905.1

View Full Product Details