Product Details

SNP ID
rs184407872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:137493915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCGTTCGCGTCCGTTGATCTTCA[G/T]CCTCAACTGCTGTAGGTCCTGCTCC
Phenotype
MIM: 609323
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OLIG3 PubMed Links

Gene Details

Gene
OLIG3
Gene Name
oligodendrocyte transcription factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175747.2 480 Missense Mutation ATG,CTG M86L NP_786923.1

View Full Product Details