Product Details
- SNP ID
-
hCV179745688
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:130398980 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCCGCCACCTGGCAGATTCTGAG[A/C]GCTTCGGGCACCAGGCACCTTGGAC
- Phenotype
-
MIM: 610523
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CEP41
PubMed Links
Gene Details
- Gene
- CEP41
- Gene Name
- centrosomal protein 41
There are no transcripts associated with this gene.
View Full Product Details