Product Details
- SNP ID
-
rs184153393
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:131052951 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTAAGTGGGCGTTCCTGGGGCCCG[A/G]TACGTGGGCACAGTGACCGGGTGGG
- Phenotype
-
MIM: 605216
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGEF4
PubMed Links
Gene Details
- Gene
- ARHGEF4
- Gene Name
- Rho guanine nucleotide exchange factor 4
There are no transcripts associated with this gene.
- Gene
- FAM168B
- Gene Name
- family with sequence similarity 168 member B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001009993.3 |
680 |
Silent Mutation |
TAC,TAT |
Y180Y |
NP_001009993.2 |
NM_001321743.1 |
680 |
Silent Mutation |
TAC,TAT |
Y180Y |
NP_001308672.1 |
NM_001321744.1 |
680 |
Silent Mutation |
TAC,TAT |
Y180Y |
NP_001308673.1 |
NM_001321745.1 |
680 |
Silent Mutation |
TAC,TAT |
Y180Y |
NP_001308674.1 |
NM_001321746.1 |
680 |
Silent Mutation |
TAC,TAT |
Y180Y |
NP_001308675.1 |
NM_001321747.1 |
680 |
Silent Mutation |
TAC,TAT |
Y180Y |
NP_001308676.1 |
NM_001321748.1 |
680 |
Silent Mutation |
TAC,TAT |
Y180Y |
NP_001308677.1 |
XM_011510594.2 |
680 |
Silent Mutation |
TAC,TAT |
Y153Y |
XP_011508896.1 |
XM_017003327.1 |
680 |
Silent Mutation |
TAC,TAT |
Y180Y |
XP_016858816.1 |
XM_017003328.1 |
680 |
Silent Mutation |
TAC,TAT |
Y153Y |
XP_016858817.1 |
- Gene
- POTEI
- Gene Name
- POTE ankyrin domain family member I
View Full Product Details