Product Details
- SNP ID
-
rs184214090
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:43873682 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTTTCAAATCAGTGTGGATTTTTC[A/G]ATGTCTAATAAAATGCTGGAAAACT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF404
PubMed Links
Gene Details
- Gene
- ZNF404
- Gene Name
- zinc finger protein 404
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001033719.2 |
532 |
Nonsense Mutation |
CGA,TGA |
R178* |
NP_001028891.2 |
View Full Product Details