Product Details

SNP ID
rs184277282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:7928234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAGGCCCATGAAAATATAATTAA[A/T]TTCATAAAAACTCCTTTATTCATAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR10A6 PubMed Links

Gene Details

Gene
OR10A6
Gene Name
olfactory receptor family 10 subfamily A member 6 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004461.1 429 Missense Mutation AAA,AAT K143N NP_001004461.1

View Full Product Details