Product Details

SNP ID
rs184292389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124626506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACATGCAAGAAAAAGAAAATGG[C/T]GGGAGGTGCTCGCAAGCTGGTTCAG
Phenotype
MIM: 608422 MIM: 610079 MIM: 610614
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PANX3 PubMed Links

Gene Details

Gene
PANX3
Gene Name
pannexin 3
There are no transcripts associated with this gene.

Gene
SIAE
Gene Name
sialic acid acetylesterase
There are no transcripts associated with this gene.

Gene
TBRG1
Gene Name
transforming growth factor beta regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032811.2 726 Missense Mutation GCG,GTG A163V NP_116200.2
XM_005271709.3 726 Intron XP_005271766.1
XM_017018442.1 726 Intron XP_016873931.1

View Full Product Details