Product Details

SNP ID
rs184019244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:91435612 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAGCCTAATCCTGCTGGCAAGA[A/G]AAAGAAGTATCTCAAGGCCGCTCTG
Phenotype
MIM: 300407
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PABPC5 PubMed Links

Gene Details

Gene
PABPC5
Gene Name
poly(A) binding protein cytoplasmic 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080832.2 475 Missense Mutation AAA,AGA K12R NP_543022.1
Gene
PABPC5-AS1
Gene Name
PABPC5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details