Product Details

SNP ID
rs184025712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:59618447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCAGCAAAGAGGTCTATCAACT[A/G]TTCAGGGGTCACCCGAGGGGACCAA
Phenotype
MIM: 606836
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GCNT3 PubMed Links

Gene Details

Gene
GCNT3
Gene Name
glucosaminyl (N-acetyl) transferase 3, mucin type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004751.2 658 Missense Mutation TAT,TGT Y70C NP_004742.1

View Full Product Details