Product Details

SNP ID
rs184037908
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5269629 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGCAGAGGGAGACGACAGGTTT[A/C]CAAAGCTGTCAAAAAATCTCTGGGT
Phenotype
MIM: 142100
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HBE1 PubMed Links

Gene Details

Gene
HBE1
Gene Name
hemoglobin subunit epsilon 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005330.3 393 Missense Mutation GGA,GTA G47V NP_005321.1

View Full Product Details