Product Details

SNP ID
rs184045774
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:189676668 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTAGCTGGTTGTCTGCCAAAGAG[A/G]ATGTGCGCTCTCAAGTAGACCTCCC
Phenotype
MIM: 609803
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKAR PubMed Links

Gene Details

Gene
ANKAR
Gene Name
ankyrin and armadillo repeat containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144708.3 752 Missense Mutation AAT,GAT N60D NP_653309.3
XM_011510673.2 752 Missense Mutation AAT,GAT N60D XP_011508975.1
XM_011510674.1 752 Missense Mutation AAT,GAT N60D XP_011508976.1
XM_011510675.2 752 Missense Mutation AAT,GAT N60D XP_011508977.1
XM_011510676.2 752 Missense Mutation AAT,GAT N60D XP_011508978.1
XM_011510677.2 752 Missense Mutation AAT,GAT N60D XP_011508979.1
XM_011510679.2 752 Missense Mutation AAT,GAT N60D XP_011508981.1
XM_011510680.1 752 Missense Mutation AAT,GAT N60D XP_011508982.1
XM_011510681.1 752 Missense Mutation AAT,GAT N60D XP_011508983.1
XM_011510682.2 752 Missense Mutation AAT,GAT N60D XP_011508984.1
XM_011510685.2 752 Intron XP_011508987.1
XM_011510686.2 752 Missense Mutation AAT,GAT N60D XP_011508988.1
XM_011510687.2 752 Intron XP_011508989.1
XM_011510688.1 752 Missense Mutation AAT,GAT N60D XP_011508990.1
XM_011510689.2 752 Missense Mutation AAT,GAT N60D XP_011508991.1
XM_011510691.2 752 Intron XP_011508993.1
XM_011510692.2 752 Intron XP_011508994.1
XM_011510693.2 752 Intron XP_011508995.1
XM_011510694.2 752 Intron XP_011508996.1
XM_017003413.1 752 Missense Mutation AAT,GAT N60D XP_016858902.1
XM_017003414.1 752 Missense Mutation AAT,GAT N60D XP_016858903.1
XM_017003415.1 752 Intron XP_016858904.1
XM_017003416.1 752 Intron XP_016858905.1
XM_017003417.1 752 Missense Mutation AAT,GAT N60D XP_016858906.1
XM_017003418.1 752 Intron XP_016858907.1
XM_017003419.1 752 Intron XP_016858908.1
XM_017003420.1 752 Intron XP_016858909.1
Gene
ASNSD1
Gene Name
asparagine synthetase domain containing 1
There are no transcripts associated with this gene.

View Full Product Details