Product Details

SNP ID
rs184886006
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50273338 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTATTTTGGATCCCGGAGAGCGA[G/T]AACCCAGACGACGACAAAATCTACT
Phenotype
MIM: 601281
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LSMEM2 PubMed Links

Gene Details

Gene
LSMEM2
Gene Name
leucine rich single-pass membrane protein 2
There are no transcripts associated with this gene.

Gene
MIR6872
Gene Name
microRNA 6872
There are no transcripts associated with this gene.

Gene
SEMA3B
Gene Name
semaphorin 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005914.2 997 Missense Mutation GAG,GAT E235D NP_001005914.1
NM_001290060.1 997 Missense Mutation GAG,GAT E235D NP_001276989.1
NM_001290061.1 997 Missense Mutation GAG,GAT E235D NP_001276990.1
NM_001290062.1 997 UTR 5 NP_001276991.1
NM_001290063.1 997 Intron NP_001276992.1
NM_004636.3 997 Missense Mutation GAG,GAT E235D NP_004627.1
Gene
SEMA3B-AS1
Gene Name
SEMA3B antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details