Product Details

SNP ID
rs184889708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:33551356 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAGCACTTTACCTCCTTATATCC[A/G]TATCGCTCACTCTGTGCCTGGTTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF122 PubMed Links

Gene Details

Gene
RNF122
Gene Name
ring finger protein 122
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024787.3 621 Silent Mutation TAC,TAT Y72Y NP_079063.2
XM_017013846.1 621 Silent Mutation TAC,TAT Y48Y XP_016869335.1

View Full Product Details