Product Details

SNP ID
rs184898622
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54191403 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTGGCGAACGGCCGCTCCCTGGT[A/G]TGGGGAGCCGAGGCGGTGCAGGCCC
Phenotype
MIM: 606048 MIM: 603631 MIM: 608754
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MBOAT7 PubMed Links

Gene Details

Gene
MBOAT7
Gene Name
membrane bound O-acyltransferase domain containing 7
There are no transcripts associated with this gene.

Gene
RPS9
Gene Name
ribosomal protein S9
There are no transcripts associated with this gene.

Gene
TSEN34
Gene Name
tRNA splicing endonuclease subunit 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077446.3 142 Silent Mutation GTA,GTG V13V NP_001070914.1
NM_001282332.1 142 Silent Mutation GTA,GTG V13V NP_001269261.1
NM_001282333.1 142 Silent Mutation GTA,GTG V16V NP_001269262.1
NM_024075.4 142 Silent Mutation GTA,GTG V13V NP_076980.2
XM_005278290.4 142 Silent Mutation GTA,GTG V16V XP_005278347.1
XM_011527294.2 142 Silent Mutation GTA,GTG V13V XP_011525596.1
XM_011527295.1 142 Silent Mutation GTA,GTG V13V XP_011525597.1

View Full Product Details