Product Details

SNP ID
rs184914659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:93784737 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCAGGAGTGGCCGGGCGCCGGGT[C/G]CAGCGCAGACCAGGACGACGAGGAA
Phenotype
MIM: 603810
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MED17 PubMed Links

Gene Details

Gene
MED17
Gene Name
mediator complex subunit 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004268.4 499 Missense Mutation TCC,TGC S75C NP_004259.3
XM_011543068.2 499 Missense Mutation TCC,TGC S75C XP_011541370.1

View Full Product Details