Product Details

SNP ID
rs184937935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:110640201 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTCTGGTCGTAGTTCTCTAACT[C/T]GCCCCCATGGCCTCTTGTTGTGTGA
Phenotype
MIM: 602452
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BUB1 PubMed Links

Gene Details

Gene
BUB1
Gene Name
BUB1 mitotic checkpoint serine/threonine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278616.1 Intron NP_001265545.1
NM_001278617.1 Intron NP_001265546.1
NM_004336.4 Intron NP_004327.1

View Full Product Details