Product Details

SNP ID
rs185031743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:73836764 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTGACCACTATGAGCCTCCCGT[C/G]CAGCCGCGCGGCCCGTGTCCCGGGT
Phenotype
MIM: 138965
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CXCL6 PubMed Links

Gene Details

Gene
CXCL6
Gene Name
C-X-C motif chemokine ligand 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002993.3 Intron NP_002984.1

View Full Product Details