Product Details

SNP ID
rs185026533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89383467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTTCCTGGACACCAAATACAAT[A/G]TGGGAATACACAACCTACTAGCCTA
Phenotype
MIM: 147690
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFIT1 PubMed Links

Gene Details

Gene
IFIT1
Gene Name
interferon induced protein with tetratricopeptide repeats 1
There are no transcripts associated with this gene.

Gene
IFIT1B
Gene Name
interferon induced protein with tetratricopeptide repeats 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010987.2 234 Missense Mutation ATG,GTG M52V NP_001010987.1

View Full Product Details