Product Details
- SNP ID
-
rs185010819
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:126572459 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCAGCAAGCCAGCCCCACACTCA[A/G]TCGTGCCCTCACAGGCTCCAGTGGC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TXNRD3NB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73195848] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TXNRD3NB
- Gene Name
- thioredoxin reductase 3 neighbor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001039783.1 |
564 |
Silent Mutation |
CTG,TTG |
L29L |
NP_001034872.1 |
View Full Product Details