Product Details

SNP ID
rs184728647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:133121652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGATCCAATGGGGAGATCACAGT[A/G]AGCCTGTACAACTGATATTCCACGG
Phenotype
MIM: 194538
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF10 PubMed Links

Gene Details

Gene
ZNF10
Gene Name
zinc finger protein 10
There are no transcripts associated with this gene.

Gene
ZNF891
Gene Name
zinc finger protein 891
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277291.1 458 Silent Mutation CTC,CTT L89L NP_001264220.1
XM_017018666.1 458 Silent Mutation CTC,CTT L89L XP_016874155.1
XM_017018667.1 458 Silent Mutation CTC,CTT L89L XP_016874156.1
XM_017018668.1 458 Silent Mutation CTC,CTT L89L XP_016874157.1

View Full Product Details