Product Details

SNP ID
rs184697812
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:134234989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGATCACCCAGTCGCAGGCCTTC[A/G]ACTTTGAGTTTACCCACGTGGAGAA
Phenotype
MIM: 609276 MIM: 610027
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NCAPD3 PubMed Links

Gene Details

Gene
NCAPD3
Gene Name
non-SMC condensin II complex subunit D3
There are no transcripts associated with this gene.

Gene
VPS26B
Gene Name
VPS26, retromer complex component B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052875.4 834 Missense Mutation AAC,GAC N106D NP_443107.1
XM_011542565.2 834 Missense Mutation AAC,GAC N106D XP_011540867.1

View Full Product Details