Product Details

SNP ID
rs184667517
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:136976586 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGATCTAAAGTGAGATTTTACAA[G/T]GTCGTGATTTTTAATATACTTCATA
Phenotype
MIM: 602264
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SPOCK1 PubMed Links

Gene Details

Gene
SPOCK1
Gene Name
sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004598.3 3539 UTR 3 NP_004589.1

View Full Product Details