Product Details

SNP ID
rs184749
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:916737 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCAGTAATAATAATAATAATAAT[A/G]AGGCTGTGGCGTTTGCAAGGTGCCC
Phenotype
MIM: 603265 MIM: 604161
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARID3A PubMed Links

Gene Details

Gene
ARID3A
Gene Name
AT-rich interaction domain 3A
There are no transcripts associated with this gene.

Gene
KISS1R
Gene Name
KISS1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032551.4 45 Intron NP_115940.2
XM_017027382.1 45 UTR 5 XP_016882871.1
Gene
R3HDM4
Gene Name
R3H domain containing 4
There are no transcripts associated with this gene.

View Full Product Details