Product Details

SNP ID
rs184794884
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5393530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCATACATGCCTCAGAAACTTTGC[A/G]CCACTTCCTCATAGTCCCTCTCCAG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TUBAL3 PubMed Links

Gene Details

Gene
TUBAL3
Gene Name
tubulin alpha like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171864.1 1366 Missense Mutation GCG,GTG A403V NP_001165335.1
NM_024803.2 1366 Missense Mutation GCG,GTG A443V NP_079079.1

View Full Product Details