Product Details

SNP ID
rs184804713
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:200853989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCGATCGTGGTGAAAATATTTAC[A/G]TTTCCTAAAAATAAGTCAATATCCA
Phenotype
MIM: 601951
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLK1 PubMed Links

Gene Details

Gene
CLK1
Gene Name
CDC like kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162407.1 1406 Missense Mutation CGT,TGT R451C NP_001155879.1
NM_004071.3 1406 Missense Mutation CGT,TGT R409C NP_004062.2

View Full Product Details