Product Details
- SNP ID
-
rs184852492
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:69355835 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAATGAAATTAGGCATTTATATTC[A/C]ATCGGATTTTTTTTAAGCTTTAAAA
- Phenotype
-
MIM: 602027
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TERF2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139966848] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TERF2
- Gene Name
- telomeric repeat binding factor 2
- Gene
- TMED6
- Gene Name
- transmembrane p24 trafficking protein 6
There are no transcripts associated with this gene.
View Full Product Details