Product Details

SNP ID
rs184595966
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:173041522 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGGATTTTAGATTTGTTTGTTA[A/G]AGTTTGTATCATGTCTTTGTTTTTA
Phenotype
MIM: 603898
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNFSF18 PubMed Links

Gene Details

Gene
TNFSF18
Gene Name
tumor necrosis factor superfamily member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005092.3 446 Silent Mutation CTA,TTA L149L NP_005083.2

View Full Product Details