Product Details

SNP ID
rs185438340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:18434445 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTCTGATATTAATATTTTACTG[C/T]CAGTTCAATCGTAACTGGTTTGAAA
Phenotype
MIM: 300203
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDKL5 PubMed Links

Gene Details

Gene
CDKL5
Gene Name
cyclin dependent kinase like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037343.1 Intron NP_001032420.1
NM_001323289.1 Intron NP_001310218.1
NM_003159.2 Intron NP_003150.1

View Full Product Details