Product Details

SNP ID
rs185613087
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:212835995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGTAAAGGACTGATAGGCATCC[A/G]GGAAGTGCCTCTTTTGAACTCTCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPATA45 PubMed Links

Gene Details

Gene
SPATA45
Gene Name
spermatogenesis associated 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024601.2 263 Missense Mutation CCG,CTG P52L NP_001019772.1

View Full Product Details