Product Details

SNP ID
rs185244476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:67183110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATTCGAGCAAGTGCAGTCCAGG[A/G]GGTTATGACTTAAATTAATGGTGCT
Phenotype
MIM: 602226
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD180 PubMed Links

Gene Details

Gene
CD180
Gene Name
CD180 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005582.2 1890 Missense Mutation CCC,CTC P578L NP_005573.2
XM_005248504.4 1890 Missense Mutation CCC,CTC P565L XP_005248561.1

View Full Product Details