Product Details

SNP ID
rs185312781
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27247937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGCAGTTTCAGGAGAGCTCTGC[C/T]CAGGGCCTGGGCCTGAGCCTGGGGC
Phenotype
MIM: 607169
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRSS16 PubMed Links

Gene Details

Gene
PRSS16
Gene Name
protease, serine 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005865.3 141 Silent Mutation GCC,GCT A42A NP_005856.1
XM_017010161.1 141 Silent Mutation GCC,GCT A42A XP_016865650.1
XM_017010162.1 141 Silent Mutation GCC,GCT A42A XP_016865651.1
XM_017010163.1 141 Silent Mutation GCC,GCT A42A XP_016865652.1
XM_017010164.1 141 Silent Mutation GCC,GCT A42A XP_016865653.1
XM_017010165.1 141 Silent Mutation GCC,GCT A42A XP_016865654.1

View Full Product Details