Product Details

SNP ID
rs185367862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31947121 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGCTATGACGGTTACACTCTCC[A/G]GGGCTCTGCCAATCGCACCTGCCAA
Phenotype
MIM: 613927 MIM: 138470 MIM: 154040
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2 PubMed Links

Gene Details

Gene
C2
Gene Name
complement component 2
There are no transcripts associated with this gene.

Gene
C2-AS1
Gene Name
C2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
CFB
Gene Name
complement factor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001710.5 691 Missense Mutation CAG,CGG Q138R NP_001701.2
Gene
MIR1236
Gene Name
microRNA 1236
There are no transcripts associated with this gene.

Gene
NELFE
Gene Name
negative elongation factor complex member E
There are no transcripts associated with this gene.

View Full Product Details