Product Details

SNP ID
rs185383443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149436726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCAGCTCTGAGTCCTGGAAGCC[A/G]TGCTCTTCCACCTGTCCCATCAGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF777 PubMed Links

Gene Details

Gene
ZNF777
Gene Name
zinc finger protein 777
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015694.2 1361 Silent Mutation CAC,CAT H396H NP_056509.2
XM_005249980.3 1361 Silent Mutation CAC,CAT H440H XP_005250037.1
XM_011516055.2 1361 Silent Mutation CAC,CAT H396H XP_011514357.1

View Full Product Details