Product Details

SNP ID
rs185387716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30480055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCAGCAATTCATTTAATAAAAG[A/C]CAGAGAATCCGTATCCTCCATTGTA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP19-1 PubMed Links

Gene Details

Gene
KRTAP19-1
Gene Name
keratin associated protein 19-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181607.1 263 Missense Mutation GGC,GTC G88V NP_853638.1
Gene
KRTAP19-2
Gene Name
keratin associated protein 19-2
There are no transcripts associated with this gene.

View Full Product Details