Product Details
- SNP ID
-
rs185399823
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:139006230 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCTGGCTGCTGTCCCCACCAGGG[A/C]GCAGGGAAGGCGCTGGGGCTGCGGG
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PRR23A
PubMed Links
Gene Details
- Gene
- PRR23A
- Gene Name
- proline rich 23A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001134659.1 |
39 |
Silent Mutation |
GCG,GCT |
A13A |
NP_001128131.1 |
View Full Product Details