Product Details

SNP ID
rs185723182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66590502 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTGAGCTCCTGAACAGGGTTGA[C/T]TGGTGGGGATCCTTCAGGGCCGAGA
Phenotype
MIM: 603864
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC87 PubMed Links

Gene Details

Gene
CCDC87
Gene Name
coiled-coil domain containing 87
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018219.2 2582 Silent Mutation CAA,CAG Q838Q NP_060689.2
Gene
CCS
Gene Name
copper chaperone for superoxide dismutase
There are no transcripts associated with this gene.

View Full Product Details