Product Details

SNP ID
rs186026858
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:86560338 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGACCTTCTACTTGGAAAAAAAC[A/G]AAATGAATATGGACCACCAGGTAGA
Phenotype
MIM: 616857
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLCA4 PubMed Links

Gene Details

Gene
CLCA4
Gene Name
chloride channel accessory 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012128.3 472 Missense Mutation CAA,CGA Q143R NP_036260.2
XM_011541015.2 472 Missense Mutation CAA,CGA Q92R XP_011539317.1

View Full Product Details