Product Details

SNP ID
rs186105696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:70146816 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATAAAGATCCTGAGAGAAAGAGA[C/T]TCTTCAAGAGAGGTAAGCCTGGCCA
Phenotype
MIM: 300139
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGBP1 PubMed Links

Gene Details

Gene
IGBP1
Gene Name
immunoglobulin (CD79A) binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001551.2 964 Silent Mutation GAC,GAT D222D NP_001542.1
XM_017029489.1 964 Silent Mutation GAC,GAT D222D XP_016884978.1
XM_017029490.1 964 Intron XP_016884979.1

View Full Product Details