Product Details

SNP ID
rs186143987
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:247711884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCTTACTTCCTTATTACGAAGC[A/G]TATAGATGAAGGGGTTTAAAACTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR6F1 PubMed Links

Gene Details

Gene
OR6F1
Gene Name
olfactory receptor family 6 subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005286.1 872 Missense Mutation ACG,ATG T291M NP_001005286.1

View Full Product Details