Product Details

SNP ID
rs185781197
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78098000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCGTTACATCATCTCAGGAGACA[C/T]CCAGAGCCACAGCACCCTGGCTGAG
Phenotype
MIM: 605564
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CIB2 PubMed Links

Gene Details

Gene
CIB2
Gene Name
calcium and integrin binding family member 2
There are no transcripts associated with this gene.

Gene
SH2D7
Gene Name
SH2 domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101404.1 338 Missense Mutation ACC,ATC T113I NP_001094874.1
XM_011521895.2 338 Missense Mutation ACC,ATC T113I XP_011520197.1
XM_011521896.2 338 Missense Mutation ACC,ATC T80I XP_011520198.1
XM_011521899.2 338 Missense Mutation ACC,ATC T41I XP_011520201.1
XM_017022487.1 338 Missense Mutation ACC,ATC T94I XP_016877976.1
XM_017022488.1 338 UTR 5 XP_016877977.1

View Full Product Details