Product Details

SNP ID
rs185768359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57741925 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGAAGCCCCGGAGGGGCAGCTTCG[A/T]GGTGACGCTGCTGCGCCCGGACGGC
Phenotype
MIM: 607914 MIM: 616715
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
BTBD18 PubMed Links

Gene Details

Gene
BTBD18
Gene Name
BTB domain containing 18
There are no transcripts associated with this gene.

Gene
C11orf31
Gene Name
chromosome 11 open reading frame 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321335.1 585 Missense Mutation GAG,GTG E80V NP_001308264.1
NM_170746.3 585 Missense Mutation GAG,GTG E80V NP_734467.1
Gene
TMX2
Gene Name
thioredoxin related transmembrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144012.2 585 Intron NP_001137484.1
NM_015959.3 585 Intron NP_057043.1
Gene
TMX2-CTNND1
Gene Name
TMX2-CTNND1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details