Product Details

SNP ID
rs185772786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:69957583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATTTGAGGGTCAAAAAAGGGTAT[C/T]GTTGGAGATTTAAGGACAGGCATCA
Phenotype
MIM: 115460
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSN2 PubMed Links

Gene Details

Gene
CSN2
Gene Name
casein beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302770.1 382 Silent Mutation ACA,ACG T121T NP_001289699.1
NM_001891.3 382 Silent Mutation ACA,ACG T122T NP_001882.1
XM_017007760.1 382 Silent Mutation ACA,ACG T122T XP_016863249.1

View Full Product Details