Product Details

SNP ID
rs185784989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41577370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAATCTCCCGGCATCGCTGCACC[A/G]TGTCCTGAGGAGACACCACGCTGTG
Phenotype
MIM: 601786
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSDC2 PubMed Links

Gene Details

Gene
CSDC2
Gene Name
cold shock domain containing C2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014460.3 670 Intron NP_055275.1
Gene
PMM1
Gene Name
phosphomannomutase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002676.2 670 Missense Mutation ACG,ATG T246M NP_002667.2
XM_005261638.4 670 Missense Mutation ACG,ATG T149M XP_005261695.1
XM_011530229.1 670 Missense Mutation ACG,ATG T289M XP_011528531.1
XM_011530230.2 670 Missense Mutation ACG,ATG T251M XP_011528532.1
XM_011530231.2 670 Missense Mutation ACG,ATG T176M XP_011528533.1
XM_011530232.1 670 Missense Mutation ACG,ATG T149M XP_011528534.1

View Full Product Details