Product Details

SNP ID
rs185829668
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6466238 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGAACAGGTAGTGGGTGGTGATG[G/T]TCTCGGCCTCCCCAGTCTTGCTGAT
Phenotype
MIM: 609024
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
KDELR2 PubMed Links
Additional Information
For this assay, SNP(s) [rs2230263] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
KDELR2
Gene Name
KDEL endoplasmic reticulum protein retention receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100603.1 598 Intron NP_001094073.1
NM_006854.3 598 Missense Mutation AAC,ACC N146T NP_006845.1

View Full Product Details