Product Details

SNP ID
rs185838569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:155926048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCCCAACATGTCACACAAATAT[A/G]GAAGAAACGGAATCTGCAATACCTA
Phenotype
MIM: 600550 MIM: 191070
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTSO PubMed Links

Gene Details

Gene
CTSO
Gene Name
cathepsin O
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001334.2 1003 Silent Mutation TCC,TCT S318S NP_001325.1
Gene
TDO2
Gene Name
tryptophan 2,3-dioxygenase
There are no transcripts associated with this gene.

View Full Product Details